4-(Methylthio)phenylboronsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-(Methylthio)benzeneboronic acid, 4-Thioanisoleboronic acid
B23454.03
B23454.06
B23454.14
B23454.03EA
38
EUR
B23454.03
B23454.06
B23454.14
4-(Methylthio)phenylboronsäure ≥97%
4-(Methylthio)phenylboronsäure
Purity: 97%
CAS No.: 98546-51-1
Molecular Formula: C7H9BO2S
Formula Weight: 168.02
Melting Point: ca 208-212°
Beilstein Reference: 3247219
Formel:
C₇H₉BO₂S Molecular Weight: 168.02 g/mol Schmelzpunkt: ∼ 208…212 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00093410 CAS-Nummer: 98546-51-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...