6-Methoxy-2-naphthaldehyd ≥99%

Lieferant: Thermo Scientific

B23943.06 B23943.14 B23943.22
B23943.06EA 45.5 EUR
B23943.06 B23943.14 B23943.22
6-Methoxy-2-naphthaldehyd ≥99%
6-Methoxy-2-naphthaldehyd

Purity: 99% CAS No.: 3453-33-6 Molecular Formula: C12H10O2 Formula Weight: 186.21 Melting Point: 80-82° Beilstein Reference: 1945989

Formel: C₁₂H₁₀O₂
Molecular Weight: 186,21 g/mol
Schmelzpunkt: 80…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00081152
CAS-Nummer: 3453-33-6
EINECS: 222-377-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR