3-Methyl-2-thiophenboronsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Methylthiophene-2-boronic acid, (3-Methylthiophen-2-yl)boronic acid
H53201.MD
H53201.03
H53201.MDEA
65.2
EUR
H53201.MD
H53201.03
3-Methyl-2-thiophenboronsäure ≥98%
3-Methyl-2-thiopheneboronic acid
Purity: 98%
CAS No.: 177735-09-0
Molecular Formula: C5H7BO2S
Formula Weight: 142.01
Melting Point: 192-196°
MDL No.: MFCD00236049
Formel:
C₅H₇BO₂S Molecular Weight: 141.99 g/mol Schmelzpunkt: 192…196 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236049 CAS-Nummer: 177735-09-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...