2-Morpholinoanilin ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-(4-Morpholinyl)aniline

L05660.06 L05660.14
L05660.06EA 75 EUR
L05660.06 L05660.14
2-Morpholinoanilin ≥98%
2-Morpholinoanilin

Purity: 98% CAS No.: 5585-33-1 Molecular Formula: C10H14N2O Formula Weight: 178.24 Melting Point: 95-98° Beilstein Reference: 163926 UN No.: 2811 MDL No.: MFCD00047408

Formel: C₁₀H₁₄N₂O
Molecular Weight: 178,23 g/mol
Schmelzpunkt: 95…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00047408
CAS-Nummer: 5585-33-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR