2-Azepanon ≥99%

Lieferant: Thermo Scientific
Achtung

Synonyme: Azepan-2-one, 2-Oxohexamethyleneimine, 6-Aminocaproic acid lactam

L06999.22 L06999.36 L06999.0E
L06999.22EA 22.8 EUR
L06999.22 L06999.36 L06999.0E
2-Azepanon ≥99%
ε-Caprolactam

Purity: 99% CAS No.: 105-60-2 Molecular Formula: C6H11NO Formula Weight: 113.16 Flash Point: 152°(305°F) pH: 7-8.5 Melting Point: 68-72° Boiling Point: 270° Beilstein Reference: 106934 Merck Reference: 14,1761 MDL No.: MFCD00006936

Formel: C₆H₁₁NO
Molecular Weight: 113,16 g/mol
Siedepunkt: 270 °C
Schmelzpunkt: 68…72 °C
Dichte: 1,01
Flammpunkt: 152 °C (305 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006936
CAS-Nummer: 105-60-2
EINECS: 203-313-2
Merck Index: 14,1761

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR