1-Propyl-2-thioharnstoff ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
N-(n-Propyl)thiourea, 1-Propylthiourea
L10149.03
L10149.06
L10149.03EA
48.9
EUR
L10149.03
L10149.06
1-Propyl-2-thioharnstoff ≥98%
1-Propyl-2-thioharnstoff
Purity: 98%
CAS No.: 927-67-3
Molecular Formula: C4H10N2S
Formula Weight: 118.20
Odour: Odourless
Melting Point: 109-112°
Beilstein Reference: 1743064
UN No.: 2811
MDL No.: MFCD00041195
Formel:
C₄H₁₀N₂S Molecular Weight: 118,2 g/mol Schmelzpunkt: 109…112 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041195 CAS-Nummer: 927-67-3 EINECS: 213-158-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...