4-Bromphenylharnstoff ≥97%

Lieferant: Thermo Fisher Scientific

Synonyme: 1-(4-Bromophenyl)urea

L11074.03 L11074.06
L11074.03EA 27.1 EUR
L11074.03 L11074.06
4-Bromphenylharnstoff ≥97%
4-Bromphenylharnstoff

Purity: 97% CAS No.: 1967-25-5 Molecular Formula: C7H7BrN2O Formula Weight: 215.06 Beilstein Reference: 2090140 MDL No.: MFCD00025428

Formel: C₇H₇BrN₂O
Molecular Weight: 215.05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00025428
CAS-Nummer: 1967-25-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR