Benzofurazan ≥97%

Lieferant: Thermo Scientific

Synonyme: 2,1,3-Benzoxadiazole, Benzo[c][1,2,5]oxadiazole

L11229.03 L11229.06
L11229.03EA 30.1 EUR
L11229.03 L11229.06
Benzofurazan ≥97%
Benzofurazan

Purity: 97% CAS No.: 273-09-6 Molecular Formula: C6H4N2O Formula Weight: 120.11 Flash Point: 76°(169°F) Melting Point: 52-55° Boiling Point: 80°/20mm Beilstein Reference: 112261 MDL No.: MFCD00068064

Formel: C₆H₄N₂O
Molecular Weight: 120,11 g/mol
Siedepunkt: 80 °C (20 mmHg)
Schmelzpunkt: 52…55 °C
Flammpunkt: 76 °C (169 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00068064
CAS-Nummer: 273-09-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR