2-Formyl-3-thiophenboronsäure ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Formylthiophene-3-boronic acid, (2-Formylthiophen-3-yl)boronic acid
L15195.03
L15195.06
L15195.03EA
61.7
EUR
L15195.03
L15195.06
2-Formyl-3-thiophenboronsäure ≥97%
2-Formyl-3-thiophenboronsäure
Purity: 97%
CAS No.: 4347-31-3
Molecular Formula: C5H5BO3S
Formula Weight: 155.97
Storage Temperature: 2°C to 8°C
Melting Point: ca 200° dec.
Beilstein Reference: 4393584
MDL No.: MFCD01075678
Formel:
C₅H₅BO₃S Molecular Weight: 155,97 g/mol Schmelzpunkt: ∼ 200 °C (Zers.) |
MDL:
MFCD01075678 CAS-Nummer: 4347-31-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...