2-(Methylthio)phenylboronsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (2-(Methylthio)phenyl)boronic acid, 2-(Methylthio)benzeneboronic acid

L17456.03 L17456.06
L17456.03EA 26 EUR
L17456.03 L17456.06
2-(Methylthio)phenylboronsäure ≥98%
2-(Methylthio)phenylboronsäure

Purity: 98+% CAS No.: 168618-42-6 Molecular Formula: C7H9BO2S Formula Weight: 168.02 Melting Point: ca 160° Beilstein Reference: 8309529

Formel: C₇H₉BO₂S
Schmelzpunkt: ∼ 160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01318165
CAS-Nummer: 168618-42-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR