Boc-Pro-OH
Lieferant: BACHEM BIOCHEMICA
Synonyme:
N-(tert-Butoxycarbonyl)-L-proline, N-Boc-L-proline, N-α-t-Boc-L-Proline, BOC-L-Proline, (S)-1-(tert-Butoxycarbonyl)pyrrolidine-2-carboxylic acid
4000220.0100
4000220.0500
BACE4000220.0100EA
0
EUR
BACE4000220.0100
BACE4000220.0500
Boc-Pro-OH
Boc-Pro-OH
Formel:
C₁₀H₁₇NO₄ |
MDL:
MFCD00037324 CAS-Nummer: 15761-39-4 EINECS: 239-848-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...