Boc-D-Pro-OH
Lieferant: BACHEM BIOCHEMICA
Synonyme:
(2R)-1-[(tert-butoxy)carbonyl]pyrrolidine-2-carboxylic acid, N-Boc-D-proline
4000664.0005
4000664.0025
BACE4000664.0005EA
0
EUR
BACE4000664.0005
BACE4000664.0025
Boc-D-Pro-OH
Boc-D-Pro-OH
Formel:
C₁₀H₁₇NO₄ Molecular Weight: 215.25 g/mol |
MDL:
MFCD00063226 CAS-Nummer: 37784-17-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...