N-Acetyl-L-proline
Lieferant: BACHEM BIOCHEMICA
Synonyme: Ac-Pro-OH
4002573.0005 4002573.0025
BACE4002573.0005EA 0 EUR
BACE4002573.0005 BACE4002573.0025
N-Acetyl-L-proline
N-Acetyl-L-proline
Formel: C₇H₁₁NO₃
Molecular Weight: 157.169 g/mol
MDL: MFCD00020837
CAS-Nummer: 68-95-1
EINECS: 276-259-0