D(-)-Citrullin

Lieferant: BACHEM BIOCHEMICA

Synonyme: H-D-Cit-OH

4014198.0001 4014198.0005
BACE4014198.0001EA 0 EUR
BACE4014198.0001 BACE4014198.0005
D(-)-Citrullin
D(-)-Citrullin
Formel: C₆H₁₃N₃O₃
Molecular Weight: 175.19 g/mol
MDL: MFCD00216469
CAS-Nummer: 13594-51-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR