4-Chlorbenzamidoxim

Lieferant: BIONET RESEARCH

Synonyme: 4-Chloro-N'-hydroxybenzenecarboximidamide, 4-Chloro-N-hydroxybenzimidamide

10J-360S.500 10J-360S.1 10J-360S.5 10J-360S.10
BIOT10J-360S.500EA 0 EUR
BIOT10J-360S.500 BIOT10J-360S.1 BIOT10J-360S.5 BIOT10J-360S.10
4-Chlorbenzamidoxim
4-Chlorbenzamidoxim
Formel: C₇H₇ClN₂O
Molecular Weight: 170.6 g/mol
MDL: MFCD00029674
CAS-Nummer: 5033-28-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR