3-Brom-2-thiophencarbaldehyd

Lieferant: BIONET RESEARCH

Synonyme: 3-Bromothiophene-2-carboxaldehyde, 3-Bromo-2-formylthiophene

10X-0872.1 10X-0872.5 10X-0872.500
BIOT10X-0872.1EA 0 EUR
BIOT10X-0872.1 BIOT10X-0872.5 BIOT10X-0872.500
3-Brom-2-thiophencarbaldehyd
3-Brom-2-thiophencarbaldehyd
Formel: C₅H₃BrOS
Molecular Weight: 191,05 g/mol
MDL: MFCD00126680
CAS-Nummer: 930-96-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR