1-(2-Bromethoxy)-3-(trifluormethyl)benzol

Lieferant: BIONET RESEARCH

Synonyme: 3-(2-Bromoethoxy)benzotrifluoride

3F-943.500 3F-943.1 3F-943.5 3F-943.10
BIOT3F-943.500EA 0 EUR
BIOT3F-943.500 BIOT3F-943.1 BIOT3F-943.5 BIOT3F-943.10
1-(2-Bromethoxy)-3-(trifluormethyl)benzol
1-(2-Bromoethoxy)-3-(trifluoromethyl)benzene
Formel: C₉H₈BrF₃O
Molecular Weight: 269.06 g/mol
MDL: MFCD00215501
CAS-Nummer: 18800-39-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR