4'-Chloracetophenonoxim

Lieferant: BIONET RESEARCH

Synonyme: 1-(4-Chlorophenyl)-1-ethanone oxime

4F-002.500 4F-002.1 4F-002.5 4F-002.10
BIOT4F-002.500EA 0 EUR
BIOT4F-002.500 BIOT4F-002.1 BIOT4F-002.5 BIOT4F-002.10
4'-Chloracetophenonoxim
4'-Chloracetophenonoxim
Formel: C₈H₈ClNO
Molecular Weight: 169,61 g/mol
MDL: MFCD02609517
CAS-Nummer: 1956-39-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR