Acetophenonoxim

Lieferant: BIONET RESEARCH

Synonyme: Methyl phenyl ketoxime, 1-Phenyl-1-ethanone oxime, Acetophenone Oxime

6F-040.500 6F-040.1 6F-040.5
BIOT6F-040.500EA 0 EUR
BIOT6F-040.500 BIOT6F-040.1 BIOT6F-040.5
Acetophenonoxim
Acetophenonoxim
Formel: C₈H₉NO
MDL: MFCD00013931
CAS-Nummer: 613-91-2
EINECS: 210-360-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR