4-Brombenzamidoxim

Lieferant: BIONET RESEARCH

Synonyme: 4-Bromo-N-hydroxybenzimidamide

6Y-0803.10 6Y-0803.5 6Y-0803.1 6Y-0803.500
BIOT6Y-0803.10EA 0 EUR
BIOT6Y-0803.10 BIOT6Y-0803.5 BIOT6Y-0803.1 BIOT6Y-0803.500
4-Brombenzamidoxim
4-Brombenzamidoxim
Formel: C₇H₇BrN₂O
Molecular Weight: 215.05 g/mol
MDL: MFCD00019951
CAS-Nummer: 19227-14-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR