4-Brom-3-thiophencarbaldehyd

Lieferant: BIONET RESEARCH

6Y-0850.1 6Y-0850.500
BIOT6Y-0850.1EA 0 EUR
BIOT6Y-0850.1 BIOT6Y-0850.500
4-Brom-3-thiophencarbaldehyd
4-Brom-3-thiophencarbaldehyd
Formel: C₅H₃BrOS
MDL: MFCD01318960
CAS-Nummer: 18791-78-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR