N-(4-Nitrobenzoyl)morpholin
Lieferant: BIONET RESEARCH
Synonyme:
Morpholino(4-nitrophenyl)methanone
7X-0701.500
7X-0701.1
7X-0701.5
7X-0701.10
BIOT7X-0701.500EA
0
EUR
BIOT7X-0701.500
BIOT7X-0701.1
BIOT7X-0701.5
BIOT7X-0701.10
N-(4-Nitrobenzoyl)morpholin
N-(4-Nitrobenzoyl)morpholin
Formel:
C₁₁H₁₂N₂O₄ Molecular Weight: 236,23 g/mol |
MDL:
MFCD00451121 CAS-Nummer: 5397-76-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...