N-(2-Nitrobenzoyl)morpholin

Lieferant: BIONET RESEARCH

Synonyme: Morpholino(2-nitrophenyl)methanone

7X-5058.500 7X-5058.1 7X-5058.5 7X-5058.10
BIOT7X-5058.500EA 0 EUR
BIOT7X-5058.500 BIOT7X-5058.1 BIOT7X-5058.5 BIOT7X-5058.10
N-(2-Nitrobenzoyl)morpholin
N-(2-Nitrobenzoyl)morpholin
Formel: C₁₁H₁₂N₂O₄
MDL: MFCD00586005
CAS-Nummer: 26162-89-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR