N-(3-Nitrobenzoyl)morpholin

Lieferant: BIONET RESEARCH

Synonyme: Morpholino(3-nitrophenyl)methanone

7X-5087.1 7X-5087.10 7X-5087.5 7X-5087.500
BIOT7X-5087.1EA 0 EUR
BIOT7X-5087.1 BIOT7X-5087.10 BIOT7X-5087.5 BIOT7X-5087.500
N-(3-Nitrobenzoyl)morpholin
N-(3-Nitrobenzoyl)morpholin
Formel: C₁₁H₁₂N₂O₄
Molecular Weight: 236.23 g/mol
MDL: MFCD00451515
CAS-Nummer: 262162-90-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR