8-Anilino-1-naphthalinsulfonsäure

Lieferant: Cayman Chemical

Synonyme: 8-Anilinonaphthalene-1-sulphonic acid, 8-(Phenylamino)naphthalene-1-sulphonic acid

10010219-5 10010219-10 10010219-50
CAYM10010219-5EA 43 EUR
CAYM10010219-5 CAYM10010219-10 CAYM10010219-50
8-Anilino-1-naphthalinsulfonsäure
8-Anilino-1-naphthalinsulfonsäure
Formel: C₁₆H₁₃NO₃S
Molecular Weight: 299,35 g/mol
MDL: MFCD00003998
CAS-Nummer: 82-76-8
EINECS: 201-438-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR