5-Brom-2-thiophencarbaldehyd

Lieferant: CHEMPUR

Synonyme: 5-Bromo-2-thiophenecarboxaldehyde

85-0006.1 85-0006.5
CHMP85-0006.1EA 0 EUR
CHMP85-0006.1 CHMP85-0006.5
5-Brom-2-thiophencarbaldehyd
5-Brom-2-thiophencarbaldehyd
Formel: C₅H₃BrOS
Molecular Weight: 191,05 g/mol
MDL: MFCD00005432
CAS-Nummer: 4701-17-1
EINECS: 225-176-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR