2-Morpholinoanilin
Lieferant: Thermo Scientific
Synonyme:
2-(4-Morpholinyl)aniline
BTB02219DA
BTB02219EA
MAYBBTB02219DAEA
0
EUR
MAYBBTB02219DA
MAYBBTB02219EA
2-Morpholinoanilin
2-Morpholinoanilin
Formel:
C₁₀H₁₄N₂O Molecular Weight: 178,23 g/mol |
MDL:
MFCD00047408 CAS-Nummer: 5585-33-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...