4-(4-Nitrophenyl)morpholin

Lieferant: Thermo Scientific

BTB05771.1 BTB05771.10 BTB05771.50
MAYBBTB05771.1EA 0 EUR
MAYBBTB05771.1 MAYBBTB05771.10 MAYBBTB05771.50
4-(4-Nitrophenyl)morpholin
4-(4-Nitrophenyl)morpholin
Formel: C₁₀H₁₂N₂O₃
MDL: MFCD00023317
CAS-Nummer: 10389-51-2
EINECS: 233-851-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR