4'-Pentylacetophenonoxim

Lieferant: Thermo Scientific

BTB12800.1 BTB12800.10
MAYBBTB12800.1EA 0 EUR
MAYBBTB12800.1 MAYBBTB12800.10
4'-Pentylacetophenonoxim
4'-Pentylacetophenonoxim
Formel: C₁₃H₁₉NO
Molecular Weight: 205.3 g/mol
CAS-Nummer: 64128-28-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR