Methyl-3-ethoxy-2-thiophencarboxylat

Lieferant: Thermo Scientific

Synonyme: Methyl 3-ethoxythiophene-2-carboxylate

GK02484.1 GK02484.10
MAYBGK02484.1EA 0 EUR
MAYBGK02484.1 MAYBGK02484.10
Methyl-3-ethoxy-2-thiophencarboxylat
Methyl-3-ethoxy-2-thiophencarboxylat
Formel: C₈H₁₀O₃S
Molecular Weight: 186,23 g/mol
MDL: MFCD00102116
CAS-Nummer: 139926-22-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR