3-Ethoxy-2-thenoesäure

Lieferant: Thermo Scientific

Synonyme: 3-Ethoxythiophene-2-carboxylic acid

GK02486.1 GK02486.10
MAYBGK02486.1EA 0 EUR
MAYBGK02486.1 MAYBGK02486.10
3-Ethoxy-2-thenoesäure
3-Ethoxy-2-thenoesäure
Formel: C₇H₈O₃S
Molecular Weight: 172.2 g/mol
MDL: MFCD00205205
CAS-Nummer: 139926-23-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR