1-Propyl-2-thioharnstoff

Lieferant: CHEMPUR

Synonyme: N-(n-Propyl)thiourea, 1-Propylthiourea

GK03252.250 GK03252.1
CHMPGK03252.250EA 0 EUR
CHMPGK03252.250 MAYBGK03252.1
1-Propyl-2-thioharnstoff
1-Propyl-2-thioharnstoff
Formel: C₄H₁₀N₂S
Molecular Weight: 118,2 g/mol
MDL: MFCD00041195
CAS-Nummer: 927-67-3
EINECS: 213-158-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR