1,3-Dibutylharnstoff
Lieferant: CHEMPUR
JFD01108.250
JFD01108.1
CHMPJFD01108.250EA
0
EUR
CHMPJFD01108.250
MAYBJFD01108.1
1,3-Dibutylharnstoff
1,3-Dibutylharnstoff
Formel:
H₃CCH₂CH₂CH₂NHC(O)NHCH₂CH₂CH₂CH₃ Siedepunkt: 115 °C (0,03 mmHg) Schmelzpunkt: 75 °C |
MDL:
MFCD00015229 CAS-Nummer: 1792-17-2 EINECS: 217-258-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...