2-Brom-6-methoxynaphthalin

Lieferant: Thermo Scientific

SB01484.1 SB01484.10 SB01484.50
MAYBSB01484.1EA 0 EUR
MAYBSB01484.1 MAYBSB01484.10 MAYBSB01484.50
2-Brom-6-methoxynaphthalin
2-Bromo-6-methoxynaphthalene
Formel: C₁₁H₉BrO
Molecular Weight: 237,1 g/mol
MDL: MFCD00004062
CAS-Nummer: 5111-65-9
EINECS: 225-837-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR