4-Chlorbenzamidoxim

Lieferant: Thermo Scientific

Synonyme: 4-Chloro-N'-hydroxybenzenecarboximidamide, 4-Chloro-N-hydroxybenzimidamide

SPB06960DA SPB06960EA
MAYBSPB06960DAEA 0 EUR
MAYBSPB06960DA MAYBSPB06960EA
4-Chlorbenzamidoxim
4-Chlorbenzamidoxim
Formel: C₇H₇ClN₂O
Molecular Weight: 170.6 g/mol
MDL: MFCD00029674
CAS-Nummer: 5033-28-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR