4'-Propylacetophenonoxim

Lieferant: Thermo Scientific

TL00858.1 TL00858.10 TL00858.50
MAYBTL00858.1EA 0 EUR
MAYBTL00858.1 MAYBTL00858.10 MAYBTL00858.50
4'-Propylacetophenonoxim
4'-Propylacetophenonoxim
Formel: C₁₁H₁₅NO
CAS-Nummer: 64128-26-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR