Cyclobutanonoxim

Lieferant: Thermo Scientific

XBX00186.250 XBX00186.1
MAYBXBX00186.250EA 0 EUR
MAYBXBX00186.250 MAYBXBX00186.1
Cyclobutanonoxim
Cyclobutanonoxim
Formel: C₄H₇NO
Molecular Weight: 85.11 g/mol
MDL: MFCD00114552
CAS-Nummer: 2972-05-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR