2-Benzo[b]thiophenboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: Benzo[b]thiophen-2-ylboronic acid, 2-Benzothienylboronic acid

BB-2027-5G BB-2027-25G BB-2027-1G
COBBBB-2027-5GEA 76.1 EUR
COBBBB-2027-5G COBBBB-2027-25G COBBBB-2027-1G
2-Benzo[b]thiophenboronsäure
2-Benzo[b]thiophenboronsäure
Formel: C₈H₇BO₂S
Molecular Weight: 178.02 g/mol
MDL: MFCD01075674
CAS-Nummer: 98437-23-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR