3-Methyl-2-thiophenboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Methylthiophene-2-boronic acid, (3-Methylthiophen-2-yl)boronic acid
BB-2285-5G
BB-2285-1G
COBBBB-2285-5GEA
384
EUR
COBBBB-2285-5G
COBBBB-2285-1G
3-Methyl-2-thiophenboronsäure
3-Methyl-2-thiopheneboronic acid
Formel:
C₅H₇BO₂S Molecular Weight: 141,99 g/mol |
MDL:
MFCD00236049 CAS-Nummer: 177735-09-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...