2-Formyl-4-thiophenboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Formylthiophene-4-boronic acid

BB-2286-5G BB-2286-25G BB-2286-1G
COBBBB-2286-5GEA 277 EUR
COBBBB-2286-5G COBBBB-2286-25G COBBBB-2286-1G
2-Formyl-4-thiophenboronsäure
2-Formyl-4-thiophenboronsäure
Formel: C₅H₅BO₃S
Molecular Weight: 155,97 g/mol
MDL: MFCD03002365
CAS-Nummer: 175592-59-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR