3-Formyl-4-thiophenboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Formylthiophen-3-yl)boronic acid, 3-Formylthiophene-4-boronic acid

BB-2308-1G BB-2308-5G
COBBBB-2308-1GEA 193 EUR
COBBBB-2308-1G COBBBB-2308-5G
3-Formyl-4-thiophenboronsäure
3-Formyl-4-thiophenboronsäure
Formel: C₅H₅BO₃S
Molecular Weight: 155,97 g/mol
MDL: MFCD01319006
CAS-Nummer: 4347-32-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR