3-(Methylthio)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Thioanisoleboronic acid, 3-(Methylthio)benzeneboronic acid
BB-2342-5G
BB-2342-25G
BB-2342-100G
COBBBB-2342-5GEA
0
EUR
COBBBB-2342-5G
COBBBB-2342-25G
COBBBB-2342-100G
3-(Methylthio)phenylboronsäure
3-(Methylthio)phenylboronsäure
Formel:
C₇H₉BO₂S |
MDL:
MFCD01319105 CAS-Nummer: 128312-11-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...