2-(Methylthio)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-(Methylthio)phenyl)boronic acid, 2-(Methylthio)benzeneboronic acid

BB-2456-100G BB-2456-5G BB-2456-25G
COBBBB-2456-100GEA 1640 EUR
COBBBB-2456-100G COBBBB-2456-5G COBBBB-2456-25G
2-(Methylthio)phenylboronsäure
2-(Methylthio)phenylboronsäure
Formel: C₇H₉BO₂S
Molecular Weight: 168,02 g/mol
MDL: MFCD01318165
CAS-Nummer: 168618-42-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR