2-(Ethylthio)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-(Ethylthio)benzeneboronic acid

BB-2635-5G BB-2635-25G BB-2635-1G
COBBBB-2635-5GEA 345 EUR
COBBBB-2635-5G COBBBB-2635-25G COBBBB-2635-1G
2-(Ethylthio)phenylboronsäure
2-(Ethylthio)phenylboronsäure
Formel: C₈H₁₁BO₂S
Molecular Weight: 182.05 g/mol
MDL: MFCD03095256
CAS-Nummer: 362045-33-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR