5-Formyl-2-thiophenboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Formylthiophen-2-boronic acid, 5-Formylthiophene-2-boronic acid

BB-2696-5G BB-2696-25G BB-2696-1G
COBBBB-2696-5GEA 53.1 EUR
COBBBB-2696-5G COBBBB-2696-25G COBBBB-2696-1G
5-Formyl-2-thiophenboronsäure
5-Formyl-2-thiophenboronsäure
Formel: C₅H₅BO₃S
Molecular Weight: 155,97 g/mol
MDL: MFCD02093666
CAS-Nummer: 4347-33-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR