4-Methyl-3-thiophenboronsäure

Lieferant: COMBI-BLOCKS

BB-8137-1G BB-8137-5G
COBBBB-8137-1GEA 257 EUR
COBBBB-8137-1G COBBBB-8137-5G
4-Methyl-3-thiophenboronsäure
4-Methyl-3-thiopheneboronic acid
Formel: C₅H₇BO₂S
Molecular Weight: 141,99 g/mol
MDL: MFCD04039882
CAS-Nummer: 177735-11-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR