4-Methyl-2-thiophenboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Methylthiophen-2-yl)boronic acid, 4-Methylthiophene-2-boronic acid

BB-8337-1G BB-8337-25G BB-8337-5G
COBBBB-8337-1GEA 0 EUR
COBBBB-8337-1G COBBBB-8337-25G COBBBB-8337-5G
4-Methyl-2-thiophenboronsäure
4-Methyl-2-thiopheneboronic acid
Formel: C₅H₇BO₂S
Molecular Weight: 141.99 g/mol
MDL: MFCD01319040
CAS-Nummer: 162607-15-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR