3-Brom-2-thiophencarbaldehyd

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromothiophene-2-carboxaldehyde, 3-Bromo-2-formylthiophene

CA-4273-25G CA-4273-5G CA-4273-1G
COBBCA-4273-25GEA 409 EUR
COBBCA-4273-25G COBBCA-4273-5G COBBCA-4273-1G
3-Brom-2-thiophencarbaldehyd
3-Brom-2-thiophencarbaldehyd
Formel: C₅H₃BrOS
Molecular Weight: 191,05 g/mol
MDL: MFCD00126680
CAS-Nummer: 930-96-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR