N-(3-Brom-5-nitrobenzoyl)morpholin
Lieferant: COMBI-BLOCKS
Synonyme:
(3-Bromo-5-nitrophenyl)(morpholino)methanone
CA-4377-1G
CA-4377-5G
COBBCA-4377-1GEA
149
EUR
COBBCA-4377-1G
COBBCA-4377-5G
N-(3-Brom-5-nitrobenzoyl)morpholin
N-(3-Brom-5-nitrobenzoyl)morpholin
Formel:
C₁₁H₁₁BrN₂O₄ Molecular Weight: 315,12 g/mol |
MDL:
MFCD09475915 CAS-Nummer: 941294-19-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...