N-(2-Fluorbenzoyl)morpholin

Lieferant: COMBI-BLOCKS

Synonyme: (2-Fluorophenyl)(morpholino)methanone

CA-4516-5G CA-4516-25G CA-4516-1G
COBBCA-4516-5GEA 303 EUR
COBBCA-4516-5G COBBCA-4516-25G COBBCA-4516-1G
N-(2-Fluorbenzoyl)morpholin
N-(2-Fluorbenzoyl)morpholin
Formel: C₁₁H₁₂FNO₂
Molecular Weight: 209,22 g/mol
MDL: MFCD00120209
CAS-Nummer: 1978-64-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR